Gene Transcripts
- Gene Symbol
- SLC9A7
- Entrez Gene ID
- 84679
- Gene Name
- solute carrier family 9 member A7
- Gene Aliases
- NHE-7, NHE7, SLC9A6
- Chromosome Location
- Chr. X: 46599251 - 46759172 on Build GRCh38
- UniGene ID
- Hs.91389
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AL691984.1 |
- |
1 |
132 |
|
Chr. X - 46601848 - 46602383 [-] |
GenBankmRNA |
DA290146.1 |
- |
1 |
313 |
|
Chr. X - 46601986 - 46602572 [-] |
GenBankmRNA |
DA911614.1 |
- |
1 |
95 |
|
Chr. X - 46601830 - 46602354 [-] |
View Full Product Details