Gene Transcripts
- Gene Symbol
- GRAMD4
- Entrez Gene ID
- 23151
- Gene Name
- GRAM domain containing 4
- Gene Aliases
- DIP, dA59H18.1, dJ439F8.1
- Chromosome Location
- Chr. 22: 46576017 - 46682755 on Build GRCh38
- UniGene ID
- Hs.475150
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC033170.1 |
- |
1 |
195 |
|
View Full Product Details