Gene Transcripts

Gene Symbol
AKTIP
Entrez Gene ID
64400
Gene Name
AKT interacting protein
Gene Aliases
FT1, FTS
Chromosome Location
Chr. 16: 53491028 - 53504411 on Build GRCh38
UniGene ID
Hs.380897
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK098222.1 - 1 314


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB250764.1 - 1 314 Chr. 16 - 53493397 - 53493968 [-]

View Full Product Details