Gene Transcripts

Gene Symbol
SPI1
Entrez Gene ID
6688
Gene Name
Spi-1 proto-oncogene
Gene Aliases
OF, PU.1, SFPI1, SPI-1, SPI-A
Chromosome Location
Chr. 11: 47354858 - 47395640 on Build GRCh38
UniGene ID
Hs.502511
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BI519776.1 - 1 522
CD368431.1 - 2 562


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK128134.1 - 1 95 Chr. 11 - 47355335 - 47361120 [+]

View Full Product Details