Gene Transcripts
- Gene Symbol
- POPDC2
- Entrez Gene ID
- 64091
- Gene Name
- popeye domain containing 2
- Gene Aliases
- POP2
- Chromosome Location
- Chr. 3: 119642052 - 119660590 on Build GRCh38
- UniGene ID
- Hs.534383
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124602.1 |
- |
1 |
2894 |
|
View Full Product Details