Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_017021732.1 | XP_016877221.1 | 9 | 2535 | |
GenBank mRNA | AK093741.1 | - | 1 | 348 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB108808.1 | - | 1 | 348 | Chr. 14 - 50331103 - 50331667 [-] |