Gene Transcripts

Gene Symbol
PHLPP2
Entrez Gene ID
23035
Gene Name
PH domain and leucine rich repeat protein phosphatase 2
Gene Aliases
PHLPPL, PPM3B
Chromosome Location
Chr. 16: 71644924 - 71724701 on Build GRCh38
UniGene ID
Hs.709458
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK001854.1 - 9 1926
BC035267.1 - 15 2947
BX648394.1 - 2 2245

View Full Product Details