Gene Transcripts
- Gene Symbol
- DOPEY1
- Entrez Gene ID
- 23033
- Gene Name
- dopey family member 1
- Gene Aliases
- DOP1, KIAA1117, dJ202D23.2
- Chromosome Location
- Chr. 6: 83067650 - 83171351 on Build GRCh38
- UniGene ID
- Hs.520246
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC036257.1 |
- |
2, 3 |
1194 |
|
View Full Product Details