Gene Transcripts

Gene Symbol
FBRSL1
Entrez Gene ID
57666
Gene Name
fibrosin like 1
Gene Aliases
Chromosome Location
Chr. 12: 132490440 - 132585188 on Build GRCh38
UniGene ID
Hs.411138
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005266181.3 XP_005266238.1 3 6902
GenBank mRNA BC008052.2 - 1 1660

View Full Product Details