Gene Transcripts

Gene Symbol
SLC13A3
Entrez Gene ID
64849
Gene Name
solute carrier family 13 member 3
Gene Aliases
NADC3, SDCT2
Chromosome Location
Chr. 20: 46557823 - 46684485 on Build GRCh38
UniGene ID
Hs.655498
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK023307.1 - 1 794

View Full Product Details