Gene Transcripts
- Gene Symbol
- PRDM5
- Entrez Gene ID
- 11107
- Gene Name
- PR domain 5
- Gene Aliases
- BCS2, PFM2
- Chromosome Location
- Chr. 4: 120684907 - 120922866 on Build GRCh38
- UniGene ID
- Hs.666782
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC047115.1 |
- |
1 |
135 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI668592.1 |
- |
1 |
140 |
|
Chr. 4 - 120830569 - 120831254 [-] |
View Full Product Details