Gene Transcripts

Gene Symbol
PRSS12
Entrez Gene ID
8492
Gene Name
protease, serine 12
Gene Aliases
BSSP-3, BSSP3, MRT1
Chromosome Location
Chr. 4: 118280038 - 118353042 on Build GRCh38
UniGene ID
Hs.445857
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003619.3 NP_003610.2 13 4451


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BU075326.1 - 1 465 Chr. 4 - 118280035 - 118280623 [-]
GenBankmRNA BX110404.1 - 1 437 Chr. 4 - 118280035 - 118280575 [-]
GenBankmRNA BU607278.1 - 240 Chr. - - [-]

View Full Product Details