Gene Transcripts
- Gene Symbol
- ZFP92
- Entrez Gene ID
- 139735
- Gene Name
- ZFP92 zinc finger protein
- Gene Aliases
- ZNF897
- Chromosome Location
- Chr. X: 153411461 - 153426481 on Build GRCh38
- UniGene ID
- Hs.534554
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK057819.1 |
- |
1 |
172 |
|
View Full Product Details