Gene Transcripts

Gene Symbol
COA1
Entrez Gene ID
55744
Gene Name
cytochrome c oxidase assembly factor 1 homolog
Gene Aliases
C7orf44, MITRAC15
Chromosome Location
Chr. 7: 43608452 - 43729541 on Build GRCh38
UniGene ID
Hs.268887
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK127933.1 - 1 921

View Full Product Details