Gene Transcripts
- Gene Symbol
- SCN1A
- Entrez Gene ID
- 6323
- Gene Name
- sodium voltage-gated channel alpha subunit 1
- Gene Aliases
- EIEE6, FEB3, FEB3A, FHM3, GEFSP2, HBSCI, NAC1, Nav1.1, SCN1, SMEI
- Chromosome Location
- Chr. 2: 165989160 - 166149216 on Build GRCh38
- UniGene ID
- Hs.22654
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DQ993522.1 |
- |
1 |
42 |
|
View Full Product Details