Gene Transcripts

Gene Symbol
MITF
Entrez Gene ID
4286
Gene Name
melanogenesis associated transcription factor
Gene Aliases
CMM8, MI, WS2, WS2A, bHLHe32
Chromosome Location
Chr. 3: 69739435 - 69968337 on Build GRCh38
UniGene ID
Hs.166017
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017006444.1 XP_016861933.1 1 7255
GenBank mRNA AY632574.1 - 1 283

View Full Product Details