Gene Transcripts
- Gene Symbol
- PHC2
- Entrez Gene ID
- 1912
- Gene Name
- polyhomeotic homolog 2
- Gene Aliases
- EDR2, HPH2, PH2
- Chromosome Location
- Chr. 1: 33323623 - 33431017 on Build GRCh38
- UniGene ID
- Hs.524271
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055807.1 |
- |
1 |
731 |
|
View Full Product Details