Gene Transcripts
- Gene Symbol
- AHSA2
- Entrez Gene ID
- 130872
- Gene Name
- AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast)
- Gene Aliases
- Hch1
- Chromosome Location
- Chr. 2: 61177418 - 61191210 on Build GRCh38
- UniGene ID
- Hs.655602
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123709.1 |
- |
1 |
2111 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CD690496.1 |
- |
1 |
218 |
|
Chr. 2 - 61181889 - 61182399 [+] |
View Full Product Details