Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_017014678.1 | XP_016870167.1 | 16 | 5161 | |
XM_017014679.1 | XP_016870168.1 | 15 | 4888 | ||
XM_017014680.1 | XP_016870169.1 | 16 | 5161 | ||
XM_017014681.1 | XP_016870170.1 | 12 | 4069 | ||
GenBank mRNA | U88892.1 | - | 1 | 221 |