Gene Transcripts
- Gene Symbol
- TRIM22
- Entrez Gene ID
- 10346
- Gene Name
- tripartite motif containing 22
- Gene Aliases
- GPSTAF50, RNF94, STAF50
- Chromosome Location
- Chr. 11: 5689587 - 5710863 on Build GRCh38
- UniGene ID
- Hs.501778
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL360187.1 |
- |
1 |
459 |
|
View Full Product Details