Gene Transcripts

Gene Symbol
ZMIZ2
Entrez Gene ID
83637
Gene Name
zinc finger MIZ-type containing 2
Gene Aliases
NET27, TRAFIP20, ZIMP7, hZIMP7
Chromosome Location
Chr. 7: 44748547 - 44769881 on Build GRCh38
UniGene ID
Hs.600681
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK128359.1 - 1 249


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI832012.1 - 1 148 Chr. 7 - 44767049 - 44767872 [+]

View Full Product Details