Gene Transcripts
- Gene Symbol
- RFTN1
- Entrez Gene ID
- 23180
- Gene Name
- raftlin, lipid raft linker 1
- Gene Aliases
- MIG2, PIB10, PIG9, RAFTLIN
- Chromosome Location
- Chr. 3: 16315845 - 16513715 on Build GRCh38
- UniGene ID
- Hs.98910
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC041884.1 |
- |
1 |
908 |
|
View Full Product Details