Gene Transcripts
- Gene Symbol
- LOC100128950
- Entrez Gene ID
- 100128950
- Gene Name
- uncharacterized LOC100128950
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.736881
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124198.1 |
- |
1 |
454 |
|
- Gene Symbol
- PIGZ
- Entrez Gene ID
- 80235
- Gene Name
- phosphatidylinositol glycan anchor biosynthesis class Z
- Gene Aliases
- GPI-MT-IV, PIG-Z, SMP3
- Chromosome Location
- Chr. 3: 196946343 - 196969411 on Build GRCh38
- UniGene ID
- Hs.518403
- Species
- Homo sapiens
View Full Product Details