Gene Transcripts
- Gene Symbol
- Sox5
- Entrez Gene ID
- 20678
- Gene Name
- SRY (sex determining region Y)-box 5
- Gene Aliases
- A730017D01Rik, AI528773
- Chromosome Location
- Chr. 6: 143828425 - 144781989 on Build GRCm38
- UniGene ID
- Mm.1752
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK031243.1 |
- |
1 |
125 |
|
AK042703.1 |
- |
1 |
544 |
|
AK086149.1 |
- |
1 |
651 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB534494.2 |
- |
1 |
586 |
|
Chr. 6 - 143830513 - 143831102 [-] |
GenBankmRNA |
CJ290445.1 |
- |
1 |
332 |
|
Chr. 6 - 143830510 - 143830848 [-] |
View Full Product Details