Gene Transcripts
- Gene Symbol
- Mx1
- Entrez Gene ID
- 17857
- Gene Name
- MX dynamin-like GTPase 1
- Gene Aliases
- AI893580, Mx, Mx-1
- Chromosome Location
- Chr. 16: 97447035 - 97462906 on Build GRCm38
- UniGene ID
- Mm.33996
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK149635.1 |
- |
1 |
3155 |
|
AK149792.1 |
- |
1 |
3155 |
|
BC031980.1 |
- |
1 |
1081 |
|
View Full Product Details