Gene Transcripts
- Gene Symbol
- Qk
- Entrez Gene ID
- 19317
- Gene Name
- quaking
- Gene Aliases
- 1110003F05Rik, 1500005P18, QkI, l(17)-1Wis, l17Wis1
- Chromosome Location
- Chr. 17: 10202856 - 10319361 on Build GRCm38
- UniGene ID
- Mm.130694
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK003349.1 |
- |
1 |
83 |
|
AK020722.1 |
- |
1 |
636 |
|
AK168411.1 |
- |
1 |
37 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM941198.1 |
- |
1 |
351 |
|
Chr. 17 - 10204868 - 10205420 [-] |
GenBankmRNA |
BB697801.1 |
- |
1 |
294 |
|
Chr. 17 - 10204884 - 10205368 [-] |
View Full Product Details