Gene Transcripts

Gene Symbol
Poglut1
Entrez Gene ID
224143
Gene Name
protein O-glucosyltransferase 1
Gene Aliases
9630046K23Rik, Clp46, Ktelc1, Rumi
Chromosome Location
Chr. 16: 38525058 - 38550256 on Build GRCm38
UniGene ID
Mm.284366
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK076379.1 - 2026


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB007121.2 - 1 374 Chr. 16 - 38523197 - 38523859 [-]

View Full Product Details