Gene Transcripts
- Gene Symbol
- Marf1
- Entrez Gene ID
- 223989
- Gene Name
- meiosis arrest female 1
- Gene Aliases
- 4921513D23Rik, BC031575, C87306, E030019O05, Lkap, mKIAA0430
- Chromosome Location
- Chr. 16: 14109166 - 14159274 on Build GRCm38
- UniGene ID
- Mm.394072
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK076566.1 |
- |
1 |
777 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CO809720.1 |
- |
|
449 |
|
Chr. - - [-] |
GenBankmRNA |
AV254721.2 |
- |
1 |
497 |
|
Chr. 16 - 14109169 - 14109817 [-] |
GenBankmRNA |
BB041395.1 |
- |
1 |
130 |
|
Chr. 16 - 14109169 - 14109450 [-] |
GenBankmRNA |
AK085118.1 |
- |
|
766 |
|
Chr. - - [-] |
GenBankmRNA |
AV326352.1 |
- |
1 |
119 |
|
Chr. 16 - 14109171 - 14109439 [-] |
GenBankmRNA |
CA945634.1 |
- |
1 |
228 |
|
Chr. 16 - 14109173 - 14109534 [-] |
GenBankmRNA |
AV335107.1 |
- |
1 |
79 |
|
Chr. 16 - 14109178 - 14109398 [-] |
View Full Product Details