Gene Transcripts
- Gene Symbol
- Taf6l
- Entrez Gene ID
- 225895
- Gene Name
- TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor
- Gene Aliases
- 2810417N14Rik, BB223262, C530024J06Rik, Paf65a
- Chromosome Location
- Chr. 19: 8774354 - 8786417 on Build GRCm38
- UniGene ID
- Mm.220886
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK013112.1 |
- |
1 |
315 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BU758277.1 |
- |
1 |
263 |
|
Chr. 19 - 8780462 - 8781001 [-] |
View Full Product Details