Gene Transcripts
- Gene Symbol
- Rps6ka5
- Entrez Gene ID
- 73086
- Gene Name
- ribosomal protein S6 kinase, polypeptide 5
- Gene Aliases
- 3110005L17Rik, 6330404E13Rik, AI854034, MSK1, MSPK1, RLPK
- Chromosome Location
- Chr. 12: 100548433 - 100725382 on Build GRCm38
- UniGene ID
- Mm.220417
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK018114.1 |
- |
1 |
30 |
|
View Full Product Details