Gene Transcripts
- Gene Symbol
- Cdc16
- Entrez Gene ID
- 69957
- Gene Name
- CDC16 cell division cycle 16
- Gene Aliases
- 2700071J12Rik, 2810431D22Rik, APC6
- Chromosome Location
- Chr. 8: 13757646 - 13781938 on Build GRCm38
- UniGene ID
- Mm.182412
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK012511.1 |
- |
1 |
1219 |
|
View Full Product Details