Gene Transcripts
- Gene Symbol
- Ntng2
- Entrez Gene ID
- 171171
- Gene Name
- netrin G2
- Gene Aliases
- 2610016D08Rik, Lmnt2
- Chromosome Location
- Chr. 2: 29194821 - 29248040 on Build GRCm38
- UniGene ID
- Mm.442448
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK011411.1 |
- |
1 |
1060 |
|
View Full Product Details