Gene Transcripts

Gene Symbol
Krtap6-3
Entrez Gene ID
100040249
Gene Name
keratin associated protein 6-3
Gene Aliases
HGTpII.3
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.328678
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA D89901.1 - 242

View Full Product Details