Gene Transcripts
- Gene Symbol
- Cacna1c
- Entrez Gene ID
- 12288
- Gene Name
- calcium channel, voltage-dependent, L type, alpha 1C subunit
- Gene Aliases
- Cav1.2, Cchl1a1, D930026N18Rik, MBC, MELC-CC
- Chromosome Location
- Chr. 6: 118587240 - 119198921 on Build GRCm38
- UniGene ID
- Mm.41628
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK142841.1 |
- |
1 |
442 |
|
BC094579.1 |
- |
2 |
371 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY731056.1 |
- |
1 |
442 |
|
Chr. 6 - 118590877 - 118591528 [-] |
View Full Product Details