Gene Transcripts
- Gene Symbol
- Vezt
- Entrez Gene ID
- 215008
- Gene Name
- vezatin, adherens junctions transmembrane protein
- Gene Aliases
- A630071D13Rik, AI848282
- Chromosome Location
- Chr. 10: 93961522 - 94035799 on Build GRCm38
- UniGene ID
- Mm.124177
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK042209.1 |
- |
1 |
1570 |
|
View Full Product Details