Gene Transcripts
- Gene Symbol
- Lpl
- Entrez Gene ID
- 16956
- Gene Name
- lipoprotein lipase
- Gene Aliases
- Chromosome Location
- Chr. 8: 68880555 - 68906932 on Build GRCm38
- UniGene ID
- Mm.1514
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK080872.1 |
- |
1 |
286 |
|
AK154419.1 |
- |
1 |
283 |
|
AK170106.1 |
- |
1 |
114 |
|
View Full Product Details