Gene Transcripts
- Gene Symbol
- Rbbp6
- Entrez Gene ID
- 19647
- Gene Name
- retinoblastoma binding protein 6
- Gene Aliases
- 4933422O15Rik, AI316869, BB233631, C030034J04Rik, C77662, P2P-R, PACT
- Chromosome Location
- Chr. 7: 122970546 - 123002572 on Build GRCm38
- UniGene ID
- Mm.4480
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016874.1 |
- |
1 |
1354 |
|
View Full Product Details