Gene Transcripts
- Gene Symbol
- Igkv19-93
- Entrez Gene ID
- 692161
- Gene Name
- immunoglobulin kappa chain variable 19-93
- Gene Aliases
- Igk-V38, Vx38C
- Chromosome Location
- Chr. 6: 68736297 - 68736764 on Build GRCm38
- UniGene ID
- Mm.288753
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M17782.1 |
- |
2 |
305 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK157689.1 |
- |
|
339 |
|
Chr. - - [-] |
View Full Product Details