Gene Transcripts
- Gene Symbol
- Il17b
- Entrez Gene ID
- 56069
- Gene Name
- interleukin 17B
- Gene Aliases
- 1110006O16Rik, 1700006N07Rik, Zcyto7
- Chromosome Location
- Chr. 18: 61687624 - 61692538 on Build GRCm38
- UniGene ID
- Mm.59313
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK005683.1 |
- |
2 |
329 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY705861.1 |
- |
2 |
327 |
|
Chr. 18 - 61691044 - 61692384 [-] |
GenBankmRNA |
BU936273.1 |
- |
|
342 |
|
Chr. - - [-] |
View Full Product Details