Gene Transcripts

Gene Symbol
Qk
Entrez Gene ID
19317
Gene Name
quaking
Gene Aliases
1110003F05Rik, 1500005P18, QkI, l(17)-1Wis, l17Wis1
Chromosome Location
Chr. 17: 10202856 - 10319361 on Build GRCm38
UniGene ID
Mm.130694
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK033279.1 - 1 1762


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB478042.2 - 1 298 Chr. 17 - 10202858 - 10203501 [-]
GenBankmRNA AA198996.1 - 1 178 Chr. 17 - 10202921 - 10203382 [-]

View Full Product Details