Gene Transcripts
- Gene Symbol
- Son
- Entrez Gene ID
- 20658
- Gene Name
- Son DNA binding protein
- Gene Aliases
- 2900011L12Rik, AA409051, AU067731, C81487, mKIAA1019, nrebp
- Chromosome Location
- Chr. 16: 91647824 - 91679195 on Build GRCm38
- UniGene ID
- Mm.46401
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK003921.1 |
- |
|
360 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY704175.1 |
- |
1 |
360 |
|
Chr. 16 - 91679067 - 91679460 [+] |
View Full Product Details