Gene Transcripts
- Gene Symbol
- Iqcc
- Entrez Gene ID
- 230767
- Gene Name
- IQ motif containing C
- Gene Aliases
- BC026978
- Chromosome Location
- Chr. 4: 129615617 - 129619213 on Build GRCm38
- UniGene ID
- Mm.441132
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK083217.1 |
- |
1 |
156 |
|
View Full Product Details