Gene Transcripts
- Gene Symbol
- Prickle2
- Entrez Gene ID
- 243548
- Gene Name
- prickle homolog 2 (Drosophila)
- Gene Aliases
- 6230400G14Rik, 6720451F06Rik, mpk2
- Chromosome Location
- Chr. 6: 92370891 - 92706335 on Build GRCm38
- UniGene ID
- Mm.248820
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020123.1 |
- |
1 |
890 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DV063972.1 |
- |
|
257 |
|
Chr. - - [-] |
View Full Product Details