Gene Transcripts
- Gene Symbol
- Klhl3
- Entrez Gene ID
- 100503085
- Gene Name
- kelch-like 3
- Gene Aliases
- 7530408C15Rik, AI430941, EG627648
- Chromosome Location
- Chr. 13: 58004439 - 58123481 on Build GRCm38
- UniGene ID
- Mm.379256
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK033046.1 |
- |
|
3388 |
|
AK082138.1 |
- |
|
1267 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB273337.1 |
- |
1 |
274 |
|
Chr. 13 - 58000538 - 58000897 [-] |
GenBankmRNA |
AV360226.2 |
- |
1 |
273 |
|
Chr. 13 - 58000231 - 58000879 [-] |
GenBankmRNA |
AI510083.1 |
- |
1 |
429 |
|
Chr. 13 - 58000592 - 58001035 [-] |
GenBankmRNA |
BX515508.1 |
- |
1 |
441 |
|
Chr. 13 - 58000536 - 58001046 [-] |
View Full Product Details