Gene Transcripts
- Gene Symbol
- Klhl3
- Entrez Gene ID
- 100503085
- Gene Name
- kelch-like 3
- Gene Aliases
- 7530408C15Rik, AI430941, EG627648
- Chromosome Location
- Chr. 13: 58004439 - 58123481 on Build GRCm38
- UniGene ID
- Mm.379256
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK033046.1 |
- |
|
3068 |
|
AK082138.1 |
- |
|
947 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DT912221.1 |
- |
1 |
548 |
|
Chr. 13 - 58000767 - 58001476 [-] |
GenBankmRNA |
CX238931.1 |
- |
1 |
717 |
|
Chr. 13 - 58000808 - 58001643 [-] |
GenBankmRNA |
AA050068.1 |
- |
1 |
89 |
|
Chr. 13 - 58000738 - 58001015 [-] |
GenBankmRNA |
AI510083.1 |
- |
1 |
109 |
|
Chr. 13 - 58000592 - 58001035 [-] |
GenBankmRNA |
BX515508.1 |
- |
1 |
121 |
|
Chr. 13 - 58000536 - 58001046 [-] |
View Full Product Details