Gene Transcripts
- Gene Symbol
- Csmd1
- Entrez Gene ID
- 94109
- Gene Name
- CUB and Sushi multiple domains 1
- Gene Aliases
- B930082H09, mKIAA1890
- Chromosome Location
- Chr. 8: 15892542 - 17535385 on Build GRCm38
- UniGene ID
- Mm.436988
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK043289.1 |
- |
3 |
640 |
|
View Full Product Details