Gene Transcripts
- Gene Symbol
- Prune
- Entrez Gene ID
- 229589
- Gene Name
- prune homolog (Drosophila)
- Gene Aliases
- 9230112O05Rik, C130058A12, DRES-17, HTCD37, PRUNEM1, Prune-M1
- Chromosome Location
- Chr. 3: 95253674 - 95282076 on Build GRCm38
- UniGene ID
- Mm.14155
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK053118.1 |
- |
|
1057 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK082182.1 |
- |
|
1380 |
|
Chr. - - [-] |
GenBankmRNA |
CO431871.1 |
- |
1 |
176 |
|
Chr. 3 - 95281767 - 95282521 [-] |
GenBankmRNA |
BQ952404.1 |
- |
1 |
383 |
|
Chr. 3 - 95282211 - 95282728 [-] |
GenBankmRNA |
CO431624.1 |
- |
1 |
243 |
|
Chr. 3 - 95281782 - 95282588 [-] |
GenBankmRNA |
BY396907.1 |
- |
1 |
232 |
|
Chr. 3 - 95282213 - 95282577 [-] |
View Full Product Details