Gene Transcripts
- Gene Symbol
- Magea7-ps
- Entrez Gene ID
- 17143
- Gene Name
- melanoma antigen, family A, 7, pseudogene
- Gene Aliases
- Mage-a7, Magea7
- Chromosome Location
- Chr. X: 76347393 - 76347635 on Build GRCm38
- UniGene ID
- Mm.377099
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC107020.2 |
- |
1 |
122 |
|
BC127031.1 |
- |
1 |
123 |
|
BC127032.1 |
- |
1 |
122 |
|
View Full Product Details