Gene Transcripts
- Gene Symbol
- Wdr49
- Entrez Gene ID
- 213248
- Gene Name
- WD repeat domain 49
- Gene Aliases
- EG213248
- Chromosome Location
- Chr. 3: 75273928 - 75482165 on Build GRCm38
- UniGene ID
- Mm.245572
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK042714.1 |
- |
2 |
892 |
|
View Full Product Details