Gene Transcripts
- Gene Symbol
- LINC00469
- Entrez Gene ID
- 283982
- Gene Name
- long intergenic non-protein coding RNA 469
- Gene Aliases
- C17orf54
- Chromosome Location
- Chr. 17: 73749270 - 73828537 on Build GRCh38
- UniGene ID
- Hs.464079
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK097638.1 |
- |
4 |
358 |
|
BC101214.1 |
- |
4 |
459 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000321800.11 |
- |
|
459 |
|
Chr. 17 - 73750424 - 73823826 [-] |
View Full Product Details